2,4,6-Trihydroxybenzaldehyde 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A19678.03
A19678.06
A19678.14
A19678.03EA
1090
CZK
A19678.03
A19678.06
A19678.14
2,4,6-Trihydroxybenzaldehyde 95%
2,4,6-Trihydroxybenzaldehyde
Purity: 95%
CAS No.: 487-70-7
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: ca 195° dec.
Beilstein Reference: 2254429
Vzorec:
C₇H₆O₄ MW: 154.12 Bod varu: 332 Bod tání: ∼195 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00003329 CAS číslo: 487-70-7 EINECS: 207-663-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...