Salicylaldehyd ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Hydroxybenzaldehyde
A13833.0E
A13833.22
A13833.36
A13833.0EEA
9780
CZK
A13833.0E
A13833.22
A13833.36
Salicylaldehyd ≥99%
Salicylaldehyd
Purity: 99%
CAS No.: 90-02-8
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 76°(168°F)
Physical Form: Liquid
Density: 1.168
Refractive Index: 1.5730
Sensitivity: Air and Light Sensitive
Odour: Almond-like
Melting Point: -7°
Boiling Point: 197°
Beilstein Reference: 471388
Merck Reference: 14,8326
MDL No.: MFCD00003317
Precursor to a variey of chelating agents
Vzorec:
C₇H₆O₂ MW: 122,12 g/mol Bod varu: 197 °C Bod tání: –7 °C Hustota: 1,168 Bod vzplanutí: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003317 CAS číslo: 90-02-8 EINECS: 201-961-0 Merck Index: 14,8326 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...