Tisk…

3,4,5-Trimethoxybenzaldehyde ≥98%

Dodavatel: Thermo Scientific

A15038.22 A15038.14 A15038.36
A15038.22EA 2180 CZK
A15038.22 A15038.14 A15038.36
3,4,5-Trimethoxybenzaldehyde ≥98%
3,4,5-Trimethoxybenzaldehyde

Purity: 98% CAS No.: 86-81-7 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 73-77° Boiling Point: 163-165°/10mm MDL No.: MFCD00003364

Vzorec: C₁₀H₁₂O₄
MW: 196,2 g/mol
Bod varu: 163…165 °C (10 mmHg)
Bod tání: 73…77 °C
Hustota: 1,367 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003364
CAS číslo: 86-81-7
EINECS: 201-701-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR