2,4,5-Trimethoxybenzaldehyde ≥98%
Dodavatel: Thermo Fisher Scientific
B22792.14
B22792.22
B22792.14EA
1740
CZK
B22792.14
B22792.22
2,4,5-Trimethoxybenzaldehyde ≥98%
2,4,5-Trimethoxybenzaldehyde
Purity: 98%
CAS No.: 4460-86-0
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 111-115°
Boiling Point: 140°/4mm subl.
MDL No.: MFCD00003312
Vzorec:
C₁₀H₁₂O₄ Bod varu: 140 °C (4 mmHg) (subl.) Bod tání: 111…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003312 CAS číslo: 4460-86-0 EINECS: 224-713-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...