Syringaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
S7602-25G
S7602-5G
SIALS7602-25GEA
2230
CZK
SIALS7602-25G
SIALS7602-5G
Syringaldehyde, Sigma-Aldrich®
Syringaldehyde
Vzorec:
C₉H₁₀O₄ Bod varu: 322 °C (1013 hPa) Bod tání: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006943 CAS číslo: 134-96-3 EINECS: 205-167-5 Merck Index: 12,09191 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...