Tisk…

Veratraldehyde ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,4-Dimethoxybenzaldehyde

A11110.0E A11110.22 A11110.36
A11110.0EEA 12530 CZK
A11110.0E A11110.22 A11110.36
Veratraldehyde ≥99%
Veratraldehyde

Purity: 99% CAS No.: 120-14-9 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 112°(233°F) Melting Point: 42-46° Boiling Point: 281° Beilstein Reference: 473899 Merck Reference: 14,9951 MDL No.: MFCD00003363

Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Bod varu: 281 °C
Bod tání: 42…46 °C
Bod vzplanutí: 112 °C (233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS číslo: 120-14-9
EINECS: 204-373-2
Merck Index: 14,9951

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR