Tisk…

4-Heptyloxybenzaldehyde ≥97%

Dodavatel: Thermo Scientific

Synonyms: 4-(Heptyloxy)benzaldehyde

L08966.06 L08966.14
L08966.06EA 1760 CZK
L08966.06 L08966.14
4-Heptyloxybenzaldehyde ≥97%
4-Heptyloxybenzaldehyde

Purity: 97% CAS No.: 27893-41-0 Molecular Formula: C14H20O2 Formula Weight: 220.31 Refractive Index: 1.5250 Boiling Point: 152-154°/1mm MDL No.: MFCD00016616

Vzorec: C₁₄H₂₀O₂
MW: 220,31 g/mol
Bod varu: 152…154 °C (1 mmHg)
Hustota: 0,990
Storage Temperature: Ambient
MDL Number: MFCD00016616
CAS číslo: 27893-41-0
EINECS: 248-714-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR