Tisk…

4-Dodecyloxybenzaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4-(Dodecyloxy)benzaldehyde

L08889.06 L08889.03
L08889.06EA 3640 CZK
L08889.06 L08889.03
4-Dodecyloxybenzaldehyde ≥98%
4-Dodecyloxybenzaldehyde

Purity: 98% CAS No.: 24083-19-0 Molecular Formula: C19H30O2 Formula Weight: 290.45 Melting Point: 26-29° Boiling Point: 165°/0.005mm MDL No.: MFCD00043523

Vzorec: C₁₉H₃₀O₂
Bod varu: 165 °C (0,005 mmHg)
Bod tání: 26…29 °C
Storage Temperature: Ambient
MDL Number: MFCD00043523
CAS číslo: 24083-19-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR