m-Anisaldehyde 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Methoxybenzaldehyde
156760250
156761000
156765000
ACRO156760250EA
1210
CZK
ACRO156760250
ACRO156761000
ACRO156765000
m-Anisaldehyde 97%
m-Anisaldehyde
Vzorec:
C₈H₈O₂ MW: 136.15 Bod varu: 143 °C (50 mmHg) Hustota: 1.119 g/cm³ Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003361 CAS číslo: 591-31-1 EINECS: 209-712-8 |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5510 to 1.5550 (20°C, 589 nm) |
GC | ≥96.0 % |
Chloride (Cl) | ≤0.2 % |
Iodide (I) | (Limit about 0.005 %) Passes test |
Bromate (BrO3) | ≤0.002 % |
Sulfate (SO4) | ≤0.005 % |
Barium (Ba) | ≤0.002 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...