o-Anisaldehyde ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde
A10770.22
A10770.36
A10770.22EA
1570
CZK
A10770.22
A10770.36
o-Anisaldehyde ≥98%
o-Anisaldehyde
Purity: 98%
CAS No.: 135-02-4
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 117°(242°F)
Refractive Index: 1.5610
Odour: Characteristic
Melting Point: 35-39°
Boiling Point: 237-238°
Beilstein Reference: 606301
MDL No.: MFCD00003308
Vzorec:
C₈H₈O₂ MW: 136.15 Bod varu: 237…238 °C Bod tání: 35…39 °C Hustota: 1.13 Bod vzplanutí: 117 °C (242 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003308 CAS číslo: 135-02-4 EINECS: 205-171-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...