Tisk…

4-Nitrobenzaldehyde ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A11655.14 A11655.22 A11655.36
A11655.14EA 1220 CZK
A11655.14 A11655.22 A11655.36
4-Nitrobenzaldehyde ≥99%
4-Nitrobenzaldehyde

Purity: 99% CAS No.: 555-16-8 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Melting Point: 104-106° Beilstein Reference: 386796 Merck Reference: 14,6587 MDL No.: MFCD00007346

Vzorec: C₇H₅NO₃
MW: 151.12
Bod varu: 300 °C (1013 hPa)
Bod tání: 104…106 °C
Hustota: 1.546
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS číslo: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR