Tisk…

4-(Dimethylamino)benzaldehyd ACS

Dodavatel: Thermo Scientific
Varování

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

423630250 423631000 423635000 423630025
ACRO423630250EA 1340 CZK
ACRO423630250 ACRO423631000 ACRO423635000 ACRO423630025
4-(Dimethylamino)benzaldehyd ACS
4-(Dimethylamino)benzaldehyde
Vzorec: (CH₃)₂NC₆H₄CHO
MW: 149,19 g/mol
Bod varu: 176…177 °C (17 mmHg)
Bod tání: 70…75 °C
Hustota: 1,0254 g/cm³ (99.9 °C)
Bod vzplanutí: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS číslo: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Light yellow to light green Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 75 °C
Solubility (in alcohol) Passes test, (in hydrochloric acid) Passes test
Residue after ignition ≤0.1 %
Color of alcohol solution ≤60 APHA
Color of a HCl solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR