Tisk…

4-(Diethylamino)benzaldehyde 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

114001000 114005000
ACRO114001000EA 1630 CZK
ACRO114001000 ACRO114005000
4-(Diethylamino)benzaldehyde 99%
4-(Diethylamino)benzaldehyde
Vzorec: C₁₁H₁₅NO
Bod varu: 174 °C (7 mmHg)
Bod tání: 38…41 °C
Hustota: 1,03 g/cm³ (20 °C)
Bod vzplanutí: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00003382
CAS číslo: 120-21-8
EINECS: 204-377-4
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Green to brown to black Coarse crystals or chunks or crystalline solid
Infrared spectrum Conforms
Melting point 37 to 41 °C
Titration with HClO4 ≥98.5 %
Fluorescence max 605 nm (in Methanol)
Optimum lasing 614 nm (in Methanol)
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥105000 l/(mol.cm) (at Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR