Kyselina vanilinová, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Hydroxy-3-methoxybenzoic acid
M-Clarity™ quality level = MQ200
94770-10G
94770-250G
94770-50G
SIAL94770-10GEA
706
CZK
SIAL94770-10G
SIAL94770-250G
SIAL94770-50G
Kyselina vanilinová, Sigma-Aldrich®
Kyselina vanilinová
Vzorec:
C₈H₈O₄ MW: 168.15 Bod varu: 353 Storage Temperature: Ambient |
MDL Number:
MFCD00002551 CAS číslo: 121-34-6 EINECS: 204-466-8 Merck Index: 12,10068 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...