Tisk…

2,5-Dibromo-1,4-dimethoxybenzene ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1,4-Dibromo-2,5-dimethoxybenzene

A19858.09 A19858.18
A19858.09EA 4160 CZK
A19858.09 A19858.18
2,5-Dibromo-1,4-dimethoxybenzene ≥98%
2,5-Dibromo-1,4-dimethoxybenzene

Purity: 98+% CAS No.: 2674-34-2 Molecular Formula: C8H8Br2O2 Formula Weight: 295.96 Melting Point: 144-148° Beilstein Reference: 1953103

Vzorec: C₈H₈Br₂O₂
Bod tání: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00025694
CAS číslo: 2674-34-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR