Tisk…

1-Bromo-4-nitrobenzene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4-Bromonitrobenzene

A17705.06 A17705.14 A17705.22 A17705.36
A17705.06EA 886 CZK
A17705.06 A17705.14 A17705.22 A17705.36
1-Bromo-4-nitrobenzene ≥98%
1-Bromo-4-nitrobenzene

Purity: 98% CAS No.: 586-78-7 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: 124-127° Boiling Point: 254-256° UN No.: 3459 MDL No.: MFCD00007280

Vzorec: C₆H₄BrNO₂
MW: 202.01
Bod varu: 254…256 °C
Bod tání: 124…127 °C
Hustota: 1.948
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007280
CAS číslo: 586-78-7
EINECS: 209-583-8
UN: 3459
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR