2,6-Dibromoaniline ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L01285.04
L01285.09
L01285.04EA
1010
CZK
L01285.04
L01285.09
2,6-Dibromoaniline ≥97%
2,6-Dibromoaniline
Purity: 97%
CAS No.: 608-30-0
Molecular Formula: C6H5Br2N
Formula Weight: 250.93
Odour: Characteristic
Melting Point: 79-83°
Boiling Point: 262-264°
Beilstein Reference: 2207094
UN No.: 2811
MDL No.: MFCD00007638
Vzorec:
C₆H₅Br₂N MW: 250.92 Bod varu: 262…264 °C Bod tání: 79…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007638 CAS číslo: 608-30-0 EINECS: 210-158-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...