1-Bromo-4-(trifluoromethoxy)benzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Trifluoromethoxy)bromobenzene
A12104.06
A12104.14
A12104.06EA
1610
CZK
A12104.06
A12104.14
1-Bromo-4-(trifluoromethoxy)benzene ≥98%
1-Bromo-4-(trifluoromethoxy)benzene
Purity: 98%
CAS No.: 407-14-7
Molecular Formula: C7H4BrF3O
Formula Weight: 241.01
Flash Point: 67°(152°F)
Refractive Index: 1.4610
Odour: Unpleasant
Boiling Point: 155-157°
Beilstein Reference: 2046332
MDL No.: MFCD00040834
Vzorec:
C₇H₄BrF₃O MW: 241,01 g/mol Bod varu: 155…157 °C Hustota: 1,622 Bod vzplanutí: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040834 CAS číslo: 407-14-7 EINECS: 206-979-2 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...