4-Bromo-2-fluoroaniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A10228.22
A10228.09
A10228.14
A10228.22EA
3680
CZK
A10228.22
A10228.09
A10228.14
4-Bromo-2-fluoroaniline ≥98%
4-Bromo-2-fluoroaniline
Purity: 98+%
CAS No.: 367-24-8
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Flash Point: 104°(219°F)
Melting Point: 38-42°
Boiling Point: 103-108°/13mm
Beilstein Reference: 2081089
UN No.: 2811
MDL No.: MFCD00010221
Vzorec:
C₆H₅BrFN Bod varu: 103…108 °C (13 mmHg) Bod tání: 38…42 °C Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010221 CAS číslo: 367-24-8 EINECS: 206-685-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...