Methyl-o-anisate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W271705-100G-K
W271705-1KG-K
SIALW271705-100G-KEA
5300
CZK
SIALW271705-100G-K
SIALW271705-1KG-K
Methyl-o-anisate, Sigma-Aldrich®
Methyl-o-anisate
Vzorec:
C₉H₁₀O₃ |
MDL Number:
MFCD00008423 CAS číslo: 606-45-1 EINECS: 210-118-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...