Methyl-p-anisate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Methoxybenzoic acid methyl ester, 4-Anisic acid methyl ester, p-Anisic acid methyl ester, Methyl 4-methoxybenzoate
M-Clarity™ quality level = MQ300
W267902-1KG-K
W267902-5KG-K
SIALW267902-1KG-KEA
9180
CZK
SIALW267902-1KG-K
SIALW267902-5KG-K
Methyl-p-anisate, Sigma-Aldrich®
Methyl-p-anisate
Vzorec:
C₉H₁₀O₃ MW: 166,18 g/mol |
MDL Number:
MFCD00008437 CAS číslo: 121-98-2 EINECS: 204-513-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...