Ethyl pelargonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ethyl nonanoate, Pelargonic acid ethyl ester, Nonanoic acid ethyl ester
M-Clarity™ quality level = MQ300
W244708-1KG-K
W244708-20KG-K
W244708-9KG-K
SIALW244708-1KG-KEA
3540
CZK
SIALW244708-1KG-K
SIALW244708-20KG-K
SIALW244708-9KG-K
Ethyl pelargonate, Sigma-Aldrich®
Ethyl pelargonate
Vzorec:
C₁₁H₂₂O₂ MW: 186.29 |
MDL Number:
MFCD00009570 CAS číslo: 123-29-5 EINECS: 204-615-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...