Benzyl cinnamate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cinnamic acid benzyl ester, trans-Cinnamic acid benzyl ester, 3-Phenyl-2-propenoic acid benzyl ester
M-Clarity™ quality level = MQ300
W214205-1KG-K
W214205-5KG-K
SIALW214205-1KG-KEA
5010
CZK
SIALW214205-1KG-K
SIALW214205-5KG-K
Benzyl cinnamate, Sigma-Aldrich®
Benzyl cinnamate
Vzorec:
C₁₆H₁₄O₂ MW: 238.29 |
MDL Number:
MFCD00004789 CAS číslo: 103-41-3 EINECS: 203-109-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...