Methyl tricosanoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tricosanoic acid methyl ester
M-Clarity™ quality level = MQ200
T9900-100MG
T9900-1G
T9900-500MG
T9900-5G
SIALT9900-100MGEA
5210
CZK
SIALT9900-100MG
SIALT9900-1G
SIALT9900-500MG
SIALT9900-5G
Methyl tricosanoate, Sigma-Aldrich®
Methyl tricosanoate
Vzorec:
C₂₄H₄₈O₂ |
MDL Number:
MFCD00009349 CAS číslo: 2433-97-8 EINECS: 219-420-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...