Silver trifluoroacetate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
T62405-25G
T62405-5G
SIALT62405-25GEA
4240
CZK
SIALT62405-25G
SIALT62405-5G
Silver trifluoroacetate, Sigma-Aldrich®
Silver trifluoroacetate
Vzorec:
C₂AgF₃O₂ |
MDL Number:
MFCD00013199 CAS číslo: 2966-50-9 EINECS: 221-004-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...