Tisk…

4-Nitrothiophenol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Nitrobenzene-1-thiol, 4-Nitrobenzenethiol, 1-Mercapto-4-nitrobenzene, p-Nitrothiophenol

M-Clarity™ quality level = MQ100

N27209-25G N27209-5G
SIALN27209-25GEA 14020 CZK
SIALN27209-25G SIALN27209-5G
4-Nitrothiophenol, Sigma-Aldrich®
4-Nitrothiophenol
Vzorec: C₆H₅NO₂S
MW: 155.18
MDL Number: MFCD00007343
CAS číslo: 1849-36-1
EINECS: 217-436-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR