3-Nitrophenylhydrazinium chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Nitrophenylhydrazine hydrochloride, 1-(3-Nitrophenyl)hydrazine hydrochloride, (3-Nitrophenyl)hydrazine hydrochloride
M-Clarity™ quality level = MQ100
N21804-25G
N21804-5G
SIALN21804-25GEA
4770
CZK
SIALN21804-25G
SIALN21804-5G
3-Nitrophenylhydrazinium chloride, Sigma-Aldrich®
3-Nitrophenylhydrazinium chloride
Vzorec:
C₆H₈ClN₃O₂ MW: 189,6 g/mol |
MDL Number:
MFCD00012939 CAS číslo: 636-95-3 EINECS: 211-270-6 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...