Tisk…

Diethylnitrosoamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diethylnitrosamine, N-Nitrosodiethylamine

M-Clarity™ quality level = MQ100

N0756-10ML N0756-25ML
SIALN0756-10MLEA 7660 CZK
SIALN0756-10ML SIALN0756-25ML
Diethylnitrosoamine, Sigma-Aldrich®
Diethylnitrosoamine
Vzorec: C₄H₁₀N₂O
MDL Number: MFCD00013890
CAS číslo: 55-18-5
EINECS: 200-226-1
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR