Methylakrylát stabilizovaný, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Acrylic acid methyl ester
M-Clarity™ quality level = MQ200
M27301-1L
M27301-18L
M27301-250ML
M27301-5ML
SIALM27301-1LEA
2080
CZK
SIALM27301-1L
SIALM27301-18L
SIALM27301-250ML
SIALM27301-5ML
Methylakrylát stabilizovaný, Sigma-Aldrich®
Methylakrylát
Vzorec:
C₄H₆O₂ MW: 86,09 g/mol |
MDL Number:
MFCD00008627 CAS číslo: 96-33-3 EINECS: 202-500-6 UN: 1919 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...