Tisk…

2,4-Diamino-6-phenyl-1,3,5-triazine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Benzoguanamine, 2,4-Diamino-6-phenyl-s-triazine, 6-Phenyl-1,3,5-triazine-2,4-diyldiamine, 6-Phenyl-1,3,5-triazine-2,4-diamine, 2,6-Diamino-4-phenyl-1,3,5-triazine

M-Clarity™ quality level = MQ100

D23408-100G D23408-500G
SIALD23408-100GEA 695 CZK
SIALD23408-100G SIALD23408-500G
2,4-Diamino-6-phenyl-1,3,5-triazine, Sigma-Aldrich®
2,4-Diamino-6-phenyl-1,3,5-triazine
Vzorec: C₉H₉N₅
MDL Number: MFCD00023187
CAS číslo: 91-76-9
EINECS: 202-095-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR