Benzophenone hydrazone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
B9602-100G
B9602-500G
SIALB9602-100GEA
1560
CZK
SIALB9602-100G
SIALB9602-500G
Benzophenone hydrazone, Sigma-Aldrich®
Benzophenone hydrazone
Vzorec:
C₁₃H₁₂N₂ MW: 196,25 g/mol |
MDL Number:
MFCD00007624 CAS číslo: 5350-57-2 EINECS: 226-321-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...