Dapson, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
4,4'-Diaminodifenylsulfon
M-Clarity™ quality level = MQ200
A74807-100G
A74807-500G
SIALA74807-100GEA
3060
CZK
SIALA74807-100G
SIALA74807-500G
Dapson, Sigma-Aldrich®
Dapson
Vzorec:
C₁₂H₁₂N₂O₂S MW: 248,31 g/mol |
MDL Number:
MFCD00007887 CAS číslo: 80-08-0 EINECS: 201-248-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...