D(-)-Arabinóza, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
M-Clarity™ quality level = MQ200
A3131-100G
A3131-10G
A3131-25G
SIALA3131-100GEA
10150
CZK
SIALA3131-100G
SIALA3131-10G
SIALA3131-25G
D(-)-Arabinóza, Sigma-Aldrich®
D(-)-Arabinose
Vzorec:
C₅H₁₀O₅ MW: 150,13 g/mol Bod tání: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135608 CAS číslo: 10323-20-3 EINECS: 233-708-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...