Tisk…

5-Hydroxy-2-pentanone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

A20804-100G A20804-25G
SIALA20804-100GEA 9150 CZK
SIALA20804-100G SIALA20804-25G
5-Hydroxy-2-pentanone, Sigma-Aldrich®
5-Hydroxy-2-pentanone
Vzorec: C₅H₁₀O₂
Bod varu: 144 °C (1013 hPa)
Hustota: 1,01586 g/cm³ (0 °C)
MDL Number: MFCD00002961
CAS číslo: 1071-73-4
EINECS: 213-994-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR