Disodium hydrogen citrate sesquihydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
71635-250G
71635-1KG
SIAL71635-250GEA
991
CZK
SIAL71635-250G
SIAL71635-1KG
Disodium hydrogen citrate sesquihydrate, Sigma-Aldrich®
Disodium hydrogen citrate sesquihydrate
Vzorec:
C₆H₆Na₂O₇·1.5H₂O MW: 263.11 |
MDL Number:
MFCD00150445 CAS číslo: 6132-05-4 EINECS: 242-734-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...