Tisk…

Sodium dihydrogen citrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate

71498-1KG 71498-250G
SIAL71498-1KGEA 3810 CZK
SIAL71498-1KG SIAL71498-250G
Sodium dihydrogen citrate, Sigma-Aldrich®
Sodium dihydrogen citrate
Vzorec: C₆H₇NaO₇
MW: 214,11 g/mol
Bod tání: 209…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00013067
CAS číslo: 18996-35-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR