Tisk…

Methyl-5-iodo-2-methoxybenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 5-iodo-2-methoxybenzoate, 5-Iodo-2-methoxybenzoic acid methyl ester

M-Clarity™ quality level = MQ100

665304-10G 665304-1G
SIAL665304-10GEA 12760 CZK
SIAL665304-10G SIAL665304-1G
Methyl-5-iodo-2-methoxybenzoate, Sigma-Aldrich®
Methyl-5-iodo-2-methoxybenzoate
Vzorec: C₉H₉IO₃
MW: 292,07 g/mol
MDL Number: MFCD02683862
CAS číslo: 40757-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR