Tisk…

Methyl-4-amino-2-methoxybenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 4-amino-2-methoxybenzenecarboxylate, 4-Amino-2-methoxybenzoic acid methyl ester, Methyl 4-amino-2-methoxybenzoate

M-Clarity™ quality level = MQ100

647616-25G 647616-5G
SIAL647616-25GEA 3030 CZK
SIAL647616-25G SIAL647616-5G
Methyl-4-amino-2-methoxybenzoate, Sigma-Aldrich®
Methyl-4-amino-2-methoxybenzoate
Vzorec: C₉H₁₁NO₃
MW: 181,19 g/mol
MDL Number: MFCD00017202
CAS číslo: 27492-84-8
EINECS: 248-494-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR