Tisk…

Methyl-4-bromo-2-methoxybenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 4-bromo-2-methoxybenzoate, 4-Bromo-2-methoxybenzoic acid methyl ester

M-Clarity™ quality level = MQ100

647594-10G 647594-1G
SIAL647594-10GEA 4610 CZK
SIAL647594-10G SIAL647594-1G
Methyl-4-bromo-2-methoxybenzoate, Sigma-Aldrich®
Methyl-4-bromo-2-methoxybenzoate
Vzorec: C₉H₉BrO₃
MW: 245,07 g/mol
MDL Number: MFCD06797867
CAS číslo: 139102-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR