tri-Magnesium dicitrate nonahydrate, Millipore®
Dodavatel: Merck
Synonyms:
Trimagnesium Dicitrate Nonahydrate, Magnesium 2-hydroxypropane-1,2,3-tricarboxylate nonahydrate, Magnesium citrate tribasic nonahydrate
M-Clarity™ quality level = MQ200
63067-1KG-F
63067-100G-F
SIAL63067-1KG-FEA
0
CZK
SIAL63067-1KG-F
SIAL63067-100G-F
tri-Magnesium dicitrate nonahydrate, Millipore®
tri-Magnesium dicitrate nonahydrate
Vzorec:
C₁₂H₁₀Mg₃O₁₄·9H₂O MW: 613.26 |
MDL Number:
MFCD00036422 CAS číslo: 153531-96-5 EINECS: 222-093-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...