Tisk…

Potassium dihydrogen citrate ≥98%, Supelco®

Dodavatel: Merck

Synonyms: mono-Potassium citrate, Citric acid monopotassium salt, Potassium citrate monobasic

M-Clarity™ quality level = MQ100

60215-1KG 60215-250G
60215-1KGEA 5720 CZK
60215-1KG 60215-250G
Potassium dihydrogen citrate ≥98%, Supelco®
Potassium dihydrogen citrate
Vzorec: C₆H₇KO₇
MW: 230.22
MDL Number: MFCD00036472
CAS číslo: 866-83-1
EINECS: 212-753-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=422a3b9b722b275ece4ad036f679bd5b84bbf52b&vid=ebaa456566275264ebe9ce5b67e5c48944aa40b6&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

assay ≥98.0% (NT)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR