4-Bromophenyl methyl sulphone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Bromo-4-(methylsulphonyl)benzene
M-Clarity™ quality level = MQ100
556327-5G
SIAL556327-5GEA
2810
CZK
SIAL556327-5G
4-Bromophenyl methyl sulphone, Sigma-Aldrich®
4-Bromophenyl methyl sulphone
Vzorec:
C₇H₇BrO₂S MW: 235.1 |
MDL Number:
MFCD00025065 CAS číslo: 3466-32-8 EINECS: 222-421-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...