Tisk…

6-Methoxysalicylic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid

464139-10G 464139-1G
SIAL464139-10GEA 7920 CZK
SIAL464139-10G SIAL464139-1G
6-Methoxysalicylic acid, Sigma-Aldrich®
6-Methoxysalicylic acid
Vzorec: C₈H₈O₄
MW: 168,15 g/mol
MDL Number: MFCD00674090
CAS číslo: 3147-64-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR