Tisk…

(1R,2R)-(+)-1,2-Diphenylethylenediamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (1R,2R)-(+)-1,2-Diphenylethylenediamine, (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, (1R,2R)-1,2-Diphenylethane-1,2-diamine, (R,R)-DPEN, (+)-((R,R)-1,2-Diamino-2-phenylethyl)benzene

M-Clarity™ quality level = MQ100

364010-1G 364010-500MG
SIAL364010-1GEA 4720 CZK
SIAL364010-1G SIAL364010-500MG
(1R,2R)-(+)-1,2-Diphenylethylenediamine, Sigma-Aldrich®
(1R,2R)-(+)-1,2-Diphenylethylenediamine
Vzorec: C₁₄H₁₆N₂
MW: 212,29 g/mol
MDL Number: MFCD00082769
CAS číslo: 35132-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR