Disodium hydrogen citrate sesquihydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
M-Clarity™ quality level = MQ200
359084-1KG
359084-250G
SIAL359084-1KGEA
2510
CZK
SIAL359084-1KG
SIAL359084-250G
Disodium hydrogen citrate sesquihydrate, Sigma-Aldrich®
Disodium hydrogen citrate sesquihydrate
Vzorec:
C₆H₆Na₂O₇·1.5H₂O MW: 263,11 g/mol |
MDL Number:
MFCD00150445 CAS číslo: 6132-05-4 EINECS: 242-734-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...