D-Cycloserine, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
30020-1G
30020-25G
30020-5G
SIAL30020-1GEA
2890
CZK
SIAL30020-1G
SIAL30020-25G
SIAL30020-5G
D-Cycloserine, Sigma-Aldrich®
D-Cycloserine
Vzorec:
C₃H₆N₂O₂ MW: 102.09 Storage Temperature: Freezer |
MDL Number:
MFCD00005353 CAS číslo: 68-41-7 EINECS: 200-688-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...