2-Hydroxyethyl-akrylát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Hydroxyethyl acrylate, Acrylic acid 2-hydroxyethyl ester
M-Clarity™ quality level = MQ100
292818-1L
292818-250ML
292818-18L
SIAL292818-1LEA
5320
CZK
SIAL292818-1L
SIAL292818-250ML
SIAL292818-18L
2-Hydroxyethyl-akrylát, Sigma-Aldrich®
2-Hydroxyethyl-akrylát
Vzorec:
C₅H₈O₃ MW: 116,12 g/mol |
MDL Number:
MFCD00002865 CAS číslo: 818-61-1 EINECS: 212-454-9 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...