2-Ethylhexyl-akrylát, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
290815-18L
290815-1L
290815-25ML
290815-3L
SIAL290815-18LEA
17550
CZK
SIAL290815-18L
SIAL290815-1L
SIAL290815-25ML
SIAL290815-3L
2-Ethylhexyl-akrylát, Sigma-Aldrich®
2-Ethylhexyl-akrylát
Vzorec:
C₁₁H₂₀O₂ |
MDL Number:
MFCD00009495 CAS číslo: 103-11-7 EINECS: 203-080-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...