Tisk…

Methyl heptadecanoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Heptadecanoic acid methyll ester, Methyl Margarate, Margaric acid methyll ester

M-Clarity™ quality level = MQ100

286087-25G 286087-5G
SIAL286087-25GEA 20190 CZK
SIAL286087-25G SIAL286087-5G
Methyl heptadecanoate, Sigma-Aldrich®
Methyl heptadecanoate
Vzorec: C₁₈H₃₆O₂
MW: 284,48 g/mol
MDL Number: MFCD00009001
CAS číslo: 1731-92-6
EINECS: 217-055-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR