Tisk…

Toluene-p-sulphonic anhydride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride

M-Clarity™ quality level = MQ200

259764-25G 259764-5G
SIAL259764-25GEA 7430 CZK
SIAL259764-25G SIAL259764-5G
Toluene-p-sulphonic anhydride, Sigma-Aldrich®
Toluene-p-sulphonic anhydride
Vzorec: C₁₄H₁₄O₅S₂
MW: 326,39 g/mol
MDL Number: MFCD00008548
CAS číslo: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR